Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 15, 16 | 1464 | ||
NM_001286049.1 | NP_001272978.1 | 14, 15 | 1439 | |||
NM_001286058.1 | NP_001272987.1 | 13, 14 | 1366 | |||
NM_007647.3 | NP_031673.2 | 14, 15 | 1391 | |||
GenBank mRNA | AF006482.1 | - | 14, 15 | 1400 | ||
AF136571.1 | - | 14, 15 | 1425 | |||
AJ238636.1 | - | 14, 15 | 1427 | |||
AK002618.1 | - | 14, 15 | 1364 | |||
AK031581.1 | - | 15, 16 | 1444 | |||
AK036641.1 | - | 15, 16 | 1441 | |||
AK037736.1 | - | 14, 15 | 1360 | |||
AK045828.1 | - | 14, 15 | 1377 | |||
AK079267.1 | - | 13, 14 | 1288 | |||
AK080265.1 | - | 15, 16 | 1451 | |||
AK081435.1 | - | 15, 16 | 1429 | |||
AK088455.1 | - | 14, 15 | 1420 | |||
AK160950.1 | - | 12, 13 | 1285 | |||
AK163688.1 | - | 14, 15 | 1365 | |||
BC015247.1 | - | 13, 14 | 1349 | 4242560 |