Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039150.1 | NP_001034239.1 | 4 | 759 | ||
NM_001039151.1 | NP_001034240.1 | 4 | 759 | |||
NM_001177785.1 | NP_001171256.1 | 4 | 759 | |||
NM_001177786.1 | NP_001171257.1 | 4 | 759 | |||
NM_001177787.1 | NP_001171258.1 | 4 | 759 | |||
NM_009851.2 | NP_033981.2 | 4 | 759 | |||
GenBank mRNA | AK045226.1 | - | 4 | 605 | ||
AK087472.1 | - | 4 | 553 | |||
AK144125.1 | - | 4 | 562 | |||
AK152099.1 | - | 4 | 555 | |||
AK162720.1 | - | 4 | 555 | |||
AK203354.1 | - | 2 | 98 | |||
BC005676.1 | - | 4 | 538 | |||
BC051388.1 | - | 4 | 589 | 4910789 | ||
CT010324.1 | - | 4 | 440 | |||
M27129.1 | - | 4 | 466 | |||
M27130.1 | - | 4 | 472 |