Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039150.1 | NP_001034239.1 | 13 | 1845 | ||
NM_001039151.1 | NP_001034240.1 | 8 | 1200 | |||
NM_001177785.1 | NP_001171256.1 | 15 | 2076 | |||
NM_001177786.1 | NP_001171257.1 | 11 | 1599 | |||
NM_001177787.1 | NP_001171258.1 | 10 | 1497 | |||
NM_009851.2 | NP_033981.2 | 18 | 2445 | |||
GenBank mRNA | AJ251594.1 | - | 18 | 2232 | ||
AK045226.1 | - | 8 | 1046 | |||
AK144125.1 | - | 8 | 1003 | |||
AK152099.1 | - | 8 | 996 | |||
AK154411.1 | - | 8 | 991 | |||
AK162720.1 | - | 8 | 997 | |||
BC005676.1 | - | 8 | 979 | |||
BC051388.1 | - | 13 | 1675 | 4910789 | ||
CT010324.1 | - | 13 | 1526 | |||
J05163.1 | - | 8 | 862 | |||
M27129.1 | - | 8 | 907 | |||
M27130.1 | - | 8 | 913 | |||
M30655.1 | - | 8 | 981 | |||
X66081.1 | - | 10 | 1181 | |||
X66082.1 | - | 11 | 1283 | |||
X66083.1 | - | 13 | 1529 | |||
X66084.1 | - | 15 | 1760 |