Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039150.1 | NP_001034239.1 | 2 | 501 | ||
NM_001039151.1 | NP_001034240.1 | 2 | 501 | |||
NM_001177785.1 | NP_001171256.1 | 2 | 501 | |||
NM_001177786.1 | NP_001171257.1 | 2 | 501 | |||
NM_001177787.1 | NP_001171258.1 | 2 | 501 | |||
NM_009851.2 | NP_033981.2 | 2 | 501 | |||
GenBank mRNA | AJ251594.1 | - | 2 | 288 | ||
AK045226.1 | - | 2 | 347 | |||
AK087472.1 | - | 2 | 295 | |||
AK144125.1 | - | 2 | 304 | |||
AK152099.1 | - | 2 | 297 | |||
AK154411.1 | - | 2 | 292 | |||
AK162720.1 | - | 2 | 297 | |||
AK166320.1 | - | 2 | 286 | |||
BC005676.1 | - | 2 | 280 | |||
BC051388.1 | - | 2 | 331 | 4910789 | ||
CT010324.1 | - | 2 | 182 | |||
J05163.1 | - | 2 | 163 | |||
M27129.1 | - | 2 | 208 | |||
M27130.1 | - | 2 | 214 | |||
M30655.1 | - | 2 | 282 | |||
X66081.1 | - | 2 | 185 | |||
X66082.1 | - | 2 | 185 | |||
X66083.1 | - | 2 | 185 | |||
X66084.1 | - | 2 | 185 |