Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007649.4 | NP_031675.1 | 1, 2 | 120 | ||
GenBank mRNA | AB196820.1 | - | 1, 2 | 81 | ||
AB196821.1 | - | 1, 2 | 81 | |||
AB196822.1 | - | 1, 2 | 81 | |||
AB196823.1 | - | 1, 2 | 81 | |||
AK007741.1 | - | 1, 2 | 121 | |||
AK084984.1 | - | 1, 2 | 149 | |||
AK088027.1 | - | 1, 2 | 167 | |||
AK146173.1 | - | 1, 2 | 167 | |||
AK151601.1 | - | 1, 2 | 111 | |||
BC060977.1 | - | 1, 2 | 148 | 30296173 | ||
X17501.1 | - | 1, 2 | 131 | |||
X53526.1 | - | 1, 2 | 131 |