Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042580.1 | NP_001036045.1 | 5, 6 | 563 | ||
NM_001282966.1 | NP_001269895.1 | 5, 6 | 607 | |||
NM_007653.3 | NP_031679.1 | 5, 6 | 522 | |||
GenBank mRNA | AF159427.1 | - | 5, 6 | 443 | ||
AK028220.1 | - | 5, 6 | 460 | |||
AK146618.1 | - | 5, 6 | 510 | |||
AK150647.1 | - | 5, 6 | 517 | |||
AK151702.1 | - | 5, 6 | 488 | |||
AK151963.1 | - | 5, 6 | 462 | |||
AK152072.1 | - | 5, 6 | 484 | |||
AK152372.1 | - | 5, 6 | 488 | |||
AK159875.1 | - | 5, 6 | 485 | |||
AK163785.1 | - | 5, 6 | 484 | |||
AK166543.1 | - | 5, 6 | 483 | |||
AK166766.1 | - | 5, 6 | 482 | |||
AK170344.1 | - | 5, 6 | 517 | |||
AK178582.1 | - | 3, 4 | 318 | |||
AK182243.1 | - | 3, 4 | 318 | |||
AK188949.1 | - | 3, 4 | 320 | |||
AK210314.1 | - | 3, 4 | 320 | |||
AK216882.1 | - | 3, 4 | 318 | |||
BC008108.1 | - | 5, 6 | 469 | 3157993 | ||
BC012212.1 | - | 5, 6 | 469 | 3157993 | ||
BC083161.1 | - | 5, 6 | 454 | 6530532 | ||
CT010179.1 | - | 4, 5 | 418 | |||
D16432.1 | - | 5, 6 | 472 |