Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291058.1 | NP_001277987.1 | 4 | 730 | ||
GenBank mRNA | AK170443.1 | - | 4 | 654 | ||
BC021637.1 | - | 4 | 671 | 3981024 | ||
BY207885.1 | - | 3 | 221 | |||
X68273.1 | - | 4 | 713 |