Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 7 | 1002 | ||
NM_001110321.1 | NP_001103791.1 | 6 | 930 | |||
NM_001110322.1 | NP_001103792.1 | 5 | 819 | |||
NM_007654.2 | NP_031680.2 | 7 | 981 | |||
GenBank mRNA | AA203771.1 | - | 2 | 155 | ||
AF543214.1 | - | 5 | 678 | |||
AK134132.1 | - | 7 | 1005 | |||
AK144191.1 | - | 6 | 933 | |||
AK151094.1 | - | 821 | ||||
BC003824.1 | - | 7 | 939 | 3599554 | ||
J04170.1 | - | 7 | 918 | |||
S40777.1 | - | 7 | 935 |