Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252472.1 | NP_001239401.1 | 3 | 534 | ||
NM_001289470.1 | NP_001276399.1 | 3 | 534 | |||
NM_013489.3 | NP_038517.1 | 3 | 534 | |||
GenBank mRNA | AB196812.1 | - | 2 | 355 | ||
AB196813.1 | - | 2 | 355 | |||
AB196814.1 | - | 2 | 355 | |||
AB196815.1 | - | 2 | 355 | |||
AF043445.1 | - | 2 | 409 | |||
AK037385.1 | - | 2 | 388 | |||
AK040694.1 | - | 2 | 382 | |||
AK154834.1 | - | 2 | 379 | |||
AK156464.1 | - | 2 | 382 | |||
AK157075.1 | - | 2 | 382 | |||
AK171067.1 | - | 2 | 393 | |||
AK171188.1 | - | 3 | 510 | |||
BC140952.1 | - | 2 | 380 | 9055564 | ||
BC144736.1 | - | 2 | 380 | 9053284 |