Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252472.1 | NP_001239401.1 | 3 | 258 | ||
NM_001289470.1 | NP_001276399.1 | 3 | 258 | |||
NM_013489.3 | NP_038517.1 | 3 | 258 | |||
GenBank mRNA | AF043445.1 | - | 2 | 133 | ||
AK037385.1 | - | 2 | 112 | |||
AK040694.1 | - | 2 | 106 | |||
AK154834.1 | - | 2 | 103 | |||
BY205033.1 | - | 3 | 253 | |||
BY207887.1 | - | 3 | 258 |