Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019388.3 | NP_062261.3 | 4 | 779 | ||
GenBank mRNA | AF065897.1 | - | 4 | 695 | ||
AF065898.1 | - | 4 | 695 | |||
AF065899.1 | - | 4 | 695 | |||
AF065900.1 | - | 4 | 695 | |||
AK079513.1 | - | 4 | 780 | |||
AK170054.1 | - | 4 | 780 | |||
AK172548.1 | - | 4 | 782 | |||
BC013807.1 | - | 4 | 753 | 4008635 | ||
L25606.1 | - | 4 | 761 | |||
S70108.1 | - | 4 | 744 |