Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007658.3 | NP_031684.3 | 10, 11 | 1418 | ||
GenBank mRNA | AK036499.1 | - | 1 | 870 | ||
AK077609.1 | - | 10, 11 | 1036 | |||
AK077991.1 | - | 10, 11 | 1421 | |||
AK142334.1 | - | 9, 10 | 1693 | |||
BC046296.1 | - | 10, 11 | 1037 | 6486343 |