Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046840.1 | - | 1 | 1561 | ||
AK141737.1 | - | 12 | 1246 | |||
AK168287.1 | - | 12 | 1241 | |||
BC004669.1 | - | 11 | 950 | 3498846 | ||
L16926.1 | - | 11 | 1428 | |||
U15562.1 | - | 12 | 1212 |