Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016742.4 | NP_058022.1 | 7, 8 | 1025 | ||
GenBank mRNA | AK010494.1 | - | 7, 8 | 1050 | ||
AK013255.1 | - | 7, 8 | 1026 | |||
AK145671.1 | - | 7, 8 | 1057 | |||
AK168651.1 | - | 7, 8 | 1043 | |||
BC060079.1 | - | 7, 8 | 1048 | 5714770 | ||
U43076.1 | - | 7, 8 | 1063 |