Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016742.4 | NP_058022.1 | 3 | 507 | ||
GenBank mRNA | AK010494.1 | - | 3 | 532 | ||
AK013255.1 | - | 3 | 508 | |||
AK145671.1 | - | 3 | 539 | |||
AK168651.1 | - | 3 | 525 | |||
AK206081.1 | - | 1 | 149 | |||
AK213273.1 | - | 2 | 317 | |||
BC060079.1 | - | 3 | 530 | 5714770 | ||
U43076.1 | - | 3 | 545 |