Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243769.1 | NP_001230698.1 | 2 | 222 | ||
NM_009861.3 | NP_033991.1 | 2 | 222 | |||
GenBank mRNA | AK003098.1 | - | 2 | 143 | ||
AK051543.1 | - | 2 | 143 | |||
AK075567.1 | - | 2 | 141 | |||
AK144216.1 | - | 2 | 145 | |||
AK150563.1 | - | 2 | 156 | |||
AK151087.1 | - | 2 | 167 | |||
AK151726.1 | - | 2 | 144 | |||
AK153564.1 | - | 2 | 166 | |||
AK154870.1 | - | 2 | 141 | |||
AK159470.1 | - | 2 | 184 | |||
AK166281.1 | - | 2 | 141 | |||
AK167195.1 | - | 2 | 130 | |||
AK167400.1 | - | 2 | 135 | |||
AK167609.1 | - | 2 | 140 | |||
AK168013.1 | - | 1 | 135 | |||
AK168076.1 | - | 2 | 135 | |||
AK168089.1 | - | 1 | 135 | |||
AK168276.1 | - | 2 | 135 | |||
AK168503.1 | - | 2 | 156 | |||
AK168758.1 | - | 2 | 175 | |||
AK168820.1 | - | 2 | 148 | |||
AK169122.1 | - | 2 | 156 | |||
AK169232.1 | - | 2 | 135 | |||
AK169805.1 | - | 2 | 160 | |||
BC064792.1 | - | 2 | 128 | 30105331 | ||
CD551112.1 | - | 2 | 209 | |||
L11318.1 | - | 1 | 58 | |||
L78075.1 | - | 1 | 62 | |||
U37720.1 | - | 2 | 130 |