Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271566.1 | NP_001258495.1 | 11 | 1630 | ||
NM_001271567.1 | NP_001258496.1 | 10 | 1365 | |||
NM_001271568.1 | NP_001258497.1 | 10 | 1344 | |||
NM_009863.3 | NP_033993.2 | 11 | 1461 | |||
GenBank mRNA | AB018574.1 | - | 10 | 1366 | ||
AB018575.1 | - | 11 | 1462 | |||
AB018576.1 | - | 9 | 2067 | |||
AB018577.1 | - | 11 | 1432 | |||
AJ007661.1 | - | 11 | 1413 | |||
AK145921.1 | - | 11 | 1387 | |||
AK146009.1 | - | 11 | 1631 | |||
BC062149.1 | - | 10 | 2179 | |||
BC072666.1 | - | 11 | 1402 | 30611704 | ||
BC080702.1 | - | 10 | 1297 | 30474846 |