Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019707.4 | NP_062681.2 | 4, 5 | 586 | ||
GenBank mRNA | AB022100.1 | - | 4, 5 | 595 | ||
AK029195.1 | - | 4, 5 | 545 | |||
AK032409.1 | - | 4, 5 | 615 | |||
AK034995.1 | - | 4, 5 | 566 | |||
AK039438.1 | - | 4, 5 | 608 | |||
AK045201.1 | - | 4, 5 | 587 | |||
AK048724.1 | - | 4, 5 | 586 | |||
AK134649.1 | - | 4, 5 | 545 | |||
BC021628.1 | - | 4, 5 | 600 | 3987627 |