Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019753.4 | NP_062727.1 | 12 | 1552 | ||
GenBank mRNA | AF177669.1 | - | 12 | 1448 | ||
AK089791.1 | - | 12 | 1520 | |||
AK155633.1 | - | 12 | 1553 | |||
AK171733.1 | - | 12 | 1475 | |||
BC042891.1 | - | 12 | 1470 | 4208360 | ||
D87912.1 | - | 12 | 1497 |