Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111099.1 | NP_001104569.1 | 3 | 1164 | ||
NM_007669.4 | NP_031695.1 | 3 | 1138 | |||
GenBank mRNA | AK007630.1 | - | 3 | 1137 | ||
AK170467.1 | - | 3 | 1139 |