Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009875.4 | NP_034005.2 | 1, 2 | 999 | ||
GenBank mRNA | AK046676.1 | - | 1, 2 | 734 | ||
AK047669.1 | - | 1, 2 | 1000 | |||
AK050240.1 | - | 1, 2 | 986 | |||
AK158411.1 | - | 1, 2 | 979 | |||
AK162146.1 | - | 1, 2 | 719 | |||
BC014296.1 | - | 1, 2 | 982 | 4193493 | ||
CT010382.1 | - | 1, 2 | 480 | |||
U09968.1 | - | 1, 2 | 480 | |||
U10440.1 | - | 1, 2 | 697 |