Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123386.1 | NP_001116858.1 | 2 | 764 | ||
NM_009881.3 | NP_034011.1 | 5 | 714 | |||
GenBank mRNA | AF081260.1 | - | 5 | 696 | ||
AF081261.1 | - | 5 | 696 | |||
AK028162.1 | - | 2 | 460 | |||
AK156509.1 | - | 2 | 765 | |||
BC055103.1 | - | 5 | 663 | 6741468 | ||
BC062123.1 | - | 2 | 853 | 6826210 |