Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006144.3 | NP_006135.1 | 3, 4 | 394 | ||
GenBank mRNA | BC015739.1 | - | 3, 4 | 365 | 4849202 | |
BP330335.1 | - | 3, 4 | 394 | |||
CR456968.1 | - | 3, 4 | 357 | |||
M18737.1 | - | 3, 4 | 357 |