Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007683.3 | NP_031709.2 | 11 | 2050 | ||
GenBank mRNA | BC150700.1 | - | 11 | 1846 | 9087611 | |
U03113.1 | - | 11 | 1936 |