Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007684.3 | NP_031710.1 | 4 | 408 | ||
GenBank mRNA | AK003473.1 | - | 4 | 424 | ||
AK004532.1 | - | 4 | 420 | |||
AK006289.1 | - | 4 | 423 | |||
AK006891.1 | - | 4 | 408 | |||
AK011821.1 | - | 4 | 401 | |||
AK017505.1 | - | 4 | 448 | |||
BC002162.1 | - | 4 | 442 | 3485864 | ||
BC054097.1 | - | 4 | 444 | 30019209 | ||
Y12474.1 | - | 4 | 359 |