Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009888.3 | NP_034018.2 | 20 | 3413 | ||
GenBank mRNA | AK077187.1 | - | 2 | 105 | ||
AK087707.1 | - | 19 | 3163 | |||
BC066092.1 | - | 20 | 3282 | 6842070 | ||
M12660.1 | - | 20 | 3324 |