Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007686.2 | NP_031712.2 | 2 | 335 | ||
GenBank mRNA | AI326560.1 | - | 2 | 236 | ||
BC150751.1 | - | 2 | 328 | 9087662 | ||
BY773461.1 | - | 2 | 336 | |||
CJ133989.1 | - | 2 | 276 | |||
U47810.1 | - | 2 | 338 |