Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007688.2 | NP_031714.1 | 3 | 484 | ||
GenBank mRNA | AK004659.1 | - | 3 | 477 | ||
AK147174.1 | - | 3 | 398 | |||
BC007138.1 | - | 3 | 427 | 3590849 | ||
L29468.1 | - | 3 | 484 |