Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009891.2 | NP_034021.1 | 9 | 1269 | ||
GenBank mRNA | AK046406.1 | - | 9 | 1270 | ||
AK047708.1 | - | 8 | 1161 | |||
BC119322.1 | - | 8 | 1031 | 8734071 | ||
BC120589.1 | - | 8 | 1031 | 40129512 |