Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cish | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009895.3 | NP_034025.1 | 3 | 639 | ||
GenBank mRNA | AK077338.1 | - | 3 | 639 | ||
AK086040.1 | - | 228 | ||||
AK170116.1 | - | 2 | 214 | |||
AK170410.1 | - | 2 | 217 | |||
AK170554.1 | - | 3 | 440 | |||
AK170877.1 | - | 3 | 435 | |||
AK171431.1 | - | 3 | 440 | |||
BC003783.1 | - | 3 | 443 | 3487580 | ||
BC022178.1 | - | 3 | 404 | 4193243 | ||
D31943.1 | - | 3 | 475 |