Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271603.1 | NP_001258532.1 | 2 | 1201 | ||
NM_009896.2 | NP_034026.1 | 2 | 1081 | |||
GenBank mRNA | AB000677.1 | - | 1 | 925 | ||
AF120490.1 | - | 2 | 1054 | |||
AF180302.1 | - | 2 | 1059 | |||
AK028632.1 | - | 1082 | ||||
AK154706.1 | - | 1098 | ||||
U88325.1 | - | 2 | 1043 |