Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030601.3 | NP_085104.1 | 14 | 2477 | ||
GenBank mRNA | AF108501.1 | - | 14 | 2468 | ||
AF115852.1 | - | 14 | 2468 | |||
AK132498.1 | - | 14 | 2477 | |||
AK134192.1 | - | 7 | 1173 | |||
AK161052.1 | - | 14 | 2471 | |||
BC008147.1 | - | 14 | 2471 | 3583804 | ||
BC039166.1 | - | 14 | 2460 |
Gene Symbol: Clca3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047838.1 | - | 2469 | |||
AF052746.1 | - | 14 | 2498 | |||
AK032511.1 | - | 14 | 2468 | |||
AK087213.1 | - | 14 | 2470 | |||
AK137904.1 | - | 9 | 1671 | |||
AK142211.1 | - | 14 | 2470 | |||
AK161805.1 | - | 14 | 2471 | |||
BC010260.1 | - | 13 | 2606 | 3710379 | ||
BC132342.1 | - | 14 | 2472 | 40130619 | ||
BC145057.1 | - | 2472 | 9053605 |