Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013491.2 | NP_038519.1 | 20 | 2480 | ||
GenBank mRNA | AK036735.1 | - | 20 | 2482 | ||
AK161489.1 | - | 13 | 1445 | |||
AY046403.1 | - | 20 | 2305 | |||
AY046404.1 | - | 21 | 2384 | |||
BC114336.1 | - | 20 | 2445 | 40048966 |