Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013491.2 | NP_038519.1 | 11 | 1263 | ||
GenBank mRNA | AK036735.1 | - | 11 | 1265 | ||
AK161489.1 | - | 4 | 228 | |||
AY046403.1 | - | 11 | 1088 | |||
AY046404.1 | - | 12 | 1167 | |||
BC114336.1 | - | 11 | 1228 | 40048966 |