Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF124848.1 | - | 15 | 1798 | ||
BC037077.1 | - | 16 | 1709 | 5042736 |