Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016674.4 | NP_057883.1 | 3 | 644 | ||
GenBank mRNA | AF072127.1 | - | 3 | 586 | ||
AK028428.1 | - | 3 | 642 | |||
AK036762.1 | - | 3 | 616 | |||
AK036780.1 | - | 3 | 645 | |||
AK040604.1 | - | 3 | 645 | |||
AK054207.1 | - | 3 | 589 | |||
AK081601.1 | - | 3 | 642 | |||
BC002003.1 | - | 3 | 597 | 3491319 | ||
BC008536.1 | - | 3 | 545 | |||
CT010299.1 | - | 3 | 431 | |||
EU076705.1 | - | 3 | 431 |