Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042634.2 | NP_001036099.1 | 3 | 353 | ||
GenBank mRNA | AK031218.1 | - | 3 | 287 | ||
AK076199.1 | - | 3 | 288 | |||
AK135765.1 | - | 3 | 288 | |||
BC062887.1 | - | 305 | ||||
BC145905.1 | - | 3 | 256 | |||
BC145911.1 | - | 3 | 256 | |||
L29221.1 | - | 3 | 269 | |||
M38381.1 | - | 3 | 269 | |||
U11054.1 | - | 3 | 253 | |||
U21209.1 | - | 3 | 269 | |||
X57186.1 | - | 3 | 253 |