Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009906.5 | NP_034036.1 | 6 | 641 | ||
GenBank mRNA | AF111172.1 | - | 6 | 649 | ||
AJ011912.1 | - | 6 | 635 | |||
AK002418.1 | - | 6 | 660 | |||
AK048279.1 | - | 6 | 642 | |||
AK083832.1 | - | 5 | 1906 | |||
AK146759.1 | - | 6 | 634 | |||
AK155021.1 | - | 6 | 660 | |||
AK165669.1 | - | 6 | 666 | |||
AK166648.1 | - | 4 | 435 | |||
AK169924.1 | - | 6 | 648 | |||
AK170781.1 | - | 6 | 637 | |||
BC024820.1 | - | 6 | 643 | 5360558 |