Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009911.3 | NP_034041.2 | 2 | 409 | ||
GenBank mRNA | AB000803.1 | - | 2 | 390 | ||
BC031665.1 | - | 2 | 396 | 3592479 | ||
BC098322.1 | - | 2 | 386 | 2864967 | ||
U59760.1 | - | 2 | 348 | |||
X99582.1 | - | 2 | 408 | |||
Z80111.1 | - | 2 | 396 | |||
Z80112.1 | - | 1 | 313 |