Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009911.3 | NP_034041.2 | 2 | 433 | ||
GenBank mRNA | AB000803.1 | - | 2 | 414 | ||
BC031665.1 | - | 2 | 420 | 3592479 | ||
BC098322.1 | - | 2 | 410 | 2864967 | ||
U59760.1 | - | 2 | 372 | |||
X99582.1 | - | 2 | 432 | |||
Z80111.1 | - | 2 | 420 | |||
Z80112.1 | - | 1 | 337 |