Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009917.5 | NP_034047.2 | 2 | 154 | ||
GenBank mRNA | AK141906.1 | - | 2 | 125 | ||
AK154595.1 | - | 2 | 155 | |||
AK155628.1 | - | 2 | 128 | |||
AK155632.1 | - | 2 | 125 | |||
AK155867.1 | - | 2 | 119 | |||
AK170303.1 | - | 2 | 125 | |||
BC103574.1 | - | 2 | 93 | 40045779 | ||
BC103586.1 | - | 2 | 93 | 40045781 | ||
BC103587.1 | - | 2 | 93 | 40045780 | ||
BC105643.1 | - | 2 | 94 | 40045777 | ||
BY198155.1 | - | 2 | 155 | |||
D83648.1 | - | 2 | 124 | |||
U47036.1 | - | 2 | 83 | |||
X94151.1 | - | 2 | 112 |