Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009917.5 | NP_034047.2 | 2 | 925 | ||
GenBank mRNA | AK141906.1 | - | 2 | 896 | ||
AK154595.1 | - | 2 | 926 | |||
AK155628.1 | - | 2 | 899 | |||
AK155632.1 | - | 2 | 896 | |||
AK155867.1 | - | 2 | 890 | |||
AK170303.1 | - | 2 | 896 | |||
BC103574.1 | - | 2 | 864 | 40045779 | ||
BC103586.1 | - | 2 | 864 | 40045781 | ||
BC103587.1 | - | 2 | 864 | 40045780 | ||
BC105643.1 | - | 2 | 865 | 40045777 | ||
D83648.1 | - | 2 | 895 | |||
U47036.1 | - | 2 | 854 | |||
X94151.1 | - | 2 | 883 |
Set Membership: |
G-protein coupled receptor |