Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007723.2 | NP_031749.2 | 9 | 2222 | ||
GenBank mRNA | BC141261.1 | - | 9 | 1991 | 9055873 | |
M84742.1 | - | 9 | 1991 | |||
U19717.1 | - | 8 | 1957 |