Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013807.2 | NP_038835.2 | 5 | 680 | ||
GenBank mRNA | AK156754.1 | - | 5 | 692 | ||
AK209626.1 | - | 3 | 233 | |||
BC031180.1 | - | 5 | 672 | 4988360 | ||
BC063051.1 | - | 5 | 680 | 5718130 | ||
U21392.1 | - | 5 | 527 |