Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002106.3 | NP_002097.1 | 4 | 419 | ||
GenBank mRNA | AK056803.1 | - | 2 | 1027 | ||
AK315413.1 | - | 4 | 337 | |||
AY597416.1 | - | 4 | 356 | |||
BC018002.1 | - | 4 | 339 | 4271353 | ||
BC020936.1 | - | 4 | 339 | 4281642 | ||
BC103743.1 | - | 4 | 454 | 6175124 | ||
CD685635.1 | - | 4 | 452 | |||
CR457415.1 | - | 3 | 234 | |||
M37583.1 | - | 4 | 340 | |||
X52317.1 | - | 4 | 340 |