Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146318.1 | NP_001139790.1 | 1 | 918 | ||
NM_009923.2 | NP_034053.2 | 2 | 703 | |||
GenBank mRNA | AF332055.1 | - | 1 | 516 | ||
AF332056.1 | - | 1 | 516 | |||
AK041969.1 | - | 1 | 918 | |||
AK050628.1 | - | 2 | 668 | |||
AK087415.1 | - | 2 | 669 | |||
AK140280.1 | - | 1 | 919 | |||
AK154654.1 | - | 2 | 727 | |||
AK158318.1 | - | 2 | 697 | |||
AK158520.1 | - | 2 | 618 | |||
AK172318.1 | - | 2 | 668 | |||
AK172514.1 | - | 2 | 668 | |||
AK212198.1 | - | 1 | 44 | |||
BC005544.1 | - | 2 | 656 | 3585034 | ||
BC021904.1 | - | 2 | 592 | 3669076 | ||
DQ887820.1 | - | 1 | 102 | |||
M31810.1 | - | 1 | 1284 | |||
M58045.1 | - | 2 | 626 |