Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146318.1 | NP_001139790.1 | 3 | 1380 | ||
NM_009923.2 | NP_034053.2 | 4 | 1165 | |||
GenBank mRNA | AF332055.1 | - | 3 | 978 | ||
AF332056.1 | - | 3 | 978 | |||
AK041969.1 | - | 3 | 1380 | |||
AK050628.1 | - | 4 | 1130 | |||
AK087415.1 | - | 4 | 1133 | |||
AK140280.1 | - | 3 | 1381 | |||
AK154654.1 | - | 4 | 1189 | |||
AK158318.1 | - | 4 | 1159 | |||
AK158520.1 | - | 4 | 1080 | |||
AK172318.1 | - | 4 | 1128 | |||
AK172514.1 | - | 4 | 1128 | |||
BC005544.1 | - | 4 | 1118 | 3585034 | ||
BC021904.1 | - | 4 | 1054 | 3669076 | ||
M31810.1 | - | 3 | 1746 | |||
M58045.1 | - | 4 | 1088 |