Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 41 | 4572 | ||
NM_009929.3 | NP_034059.2 | 42 | 3988 | |||
GenBank mRNA | AF257775.1 | - | 4 | 514 | ||
AK014292.1 | - | 3 | 444 | |||
BC043697.1 | - | 41 | 4572 | |||
BC062931.1 | - | 42 | 3955 | |||
BC064817.1 | - | 42 | 3959 | 6332464 | ||
BC066080.1 | - | 42 | 3954 | 30355615 | ||
D17546.1 | - | 40 | 3382 | |||
L22545.1 | - | 40 | 3826 | |||
U03714.1 | - | 12 | 1408 |