Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113515.2 | NP_001106987.2 | 53 | 4372 | ||
NM_031163.3 | NP_112440.2 | 54 | 4576 | |||
GenBank mRNA | AK028295.1 | - | 28 | 2558 | ||
BC030913.1 | - | 24 | 2364 | |||
BC051383.1 | - | 53 | 4283 | 6312433 | ||
BC052326.1 | - | 53 | 4262 | 6315379 | ||
BC082331.1 | - | 54 | 4509 | 30537724 | ||
CF728108.1 | - | 2 | 67 |