Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009930.2 | NP_034060.2 | 48 | 3961 | ||
GenBank mRNA | AK019448.2 | - | 14 | 1326 | ||
AK029212.1 | - | 48 | 3867 | |||
AK029451.1 | - | 1 | 14 | |||
AK041115.1 | - | 48 | 3869 | |||
AK048546.1 | - | 48 | 3869 | |||
AK079113.1 | - | 3 | 373 | |||
AK147542.1 | - | 48 | 3867 | |||
AK157762.1 | - | 48 | 3869 | |||
AK159763.1 | - | 48 | 3870 | |||
AK159868.1 | - | 4 | 507 | |||
AK159920.1 | - | 48 | 3870 | |||
AK159985.1 | - | 48 | 3870 | |||
AK160048.1 | - | 48 | 3870 | |||
AK160107.1 | - | 48 | 3868 | |||
AK160818.1 | - | 48 | 3872 | |||
AK220558.1 | - | 48 | 3884 | |||
BC013626.1 | - | 39 | 3020 | |||
BC028248.1 | - | 39 | 3025 | |||
BC039923.1 | - | 27 | 2260 | |||
BC043089.1 | - | 48 | 3836 | 6418299 | ||
BC052398.1 | - | 48 | 3853 | 5720872 | ||
BC058724.1 | - | 48 | 3870 | 5705762 |